missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Pivalate 98.0+%, TCI America™
Supplier: TCI America P235425G
Specifications
| Potassium Pivalate | |
| 300°C | |
| C5H9KO2 | |
| 25 g | |
| WFMNHCSATCWAAQ-UHFFFAOYSA-M | |
| potassium;2,2-dimethylpropanoate | |
| 23662159 | |
| ≥98.0% (T) |
| 19455-23-3 | |
| White | |
| MFCD00671345 | |
| potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt | |
| CC(C)(C)C(=O)[O-].[K+] | |
| 140.223 | |
| 140.22 | |
| Crystalline Powder |
Chemical Identifiers
| 19455-23-3 | |
| 140.223 | |
| WFMNHCSATCWAAQ-UHFFFAOYSA-M | |
| 23662159 | |
| CC(C)(C)C(=O)[O-].[K+] |
| C5H9KO2 | |
| MFCD00671345 | |
| potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt | |
| potassium;2,2-dimethylpropanoate |
Safety and Handling
TSCA : No