accessibility menu, dialog, popup

UserName

CH3CH3H3COOK

CAS RN 19455-23-3

IUPAC Name: potassium 2,2-dimethylpropanoate
Synonyms: potassium pivalate
Molecular Weight (g/mol): 140.22
Molecular Formula: C5H9KO2
InChi Key: WFMNHCSATCWAAQ-UHFFFAOYSA-M
SMILES: [K+].CC(C)(C)C([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)