missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Pivalate 98.0+%, TCI America™
$61.51 - $180.81
Chemical Identifiers
| CAS | 19455-23-3 |
|---|---|
| Molecular Formula | C5H9KO2 |
| Molecular Weight (g/mol) | 140.223 |
| MDL Number | MFCD00671345 |
| InChI Key | WFMNHCSATCWAAQ-UHFFFAOYSA-M |
| Synonym | potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt |
| PubChem CID | 23662159 |
| IUPAC Name | potassium;2,2-dimethylpropanoate |
| SMILES | CC(C)(C)C(=O)[O-].[K+] |
Chemical Identifiers
| 19455-23-3 | |
| 140.223 | |
| WFMNHCSATCWAAQ-UHFFFAOYSA-M | |
| 23662159 | |
| CC(C)(C)C(=O)[O-].[K+] |
| C5H9KO2 | |
| MFCD00671345 | |
| potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt | |
| potassium;2,2-dimethylpropanoate |
Specifications
| 19455-23-3 | |
| White | |
| MFCD00671345 | |
| potassium pivalate, potassium trimethylacetate, pivalic acid potassium salt, potassium 2,2-dimethylpropionate, trimethylacetic acid potassium salt, 2,2-dimethylpropionic acid potassium salt | |
| CC(C)(C)C(=O)[O-].[K+] | |
| 140.223 | |
| 140.22 | |
| Crystalline Powder |
| 300°C | |
| C5H9KO2 | |
| 5 g | |
| WFMNHCSATCWAAQ-UHFFFAOYSA-M | |
| potassium;2,2-dimethylpropanoate | |
| 23662159 | |
| ≥98.0% (T) | |
| Potassium Pivalate |
Safety and Handling
TSCA : No