missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Monobasic (Crystalline/NF/FCC), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLP380212

 View more versions of this product

Catalog No. P380-212

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Phosphate Monobasic
7778-77-0
4.5
2.5 kg
MFCD00011401 MFCD00147253
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate
516951
136.09
FCC/NF
1.0% max. (NF/FCC)
>250°C
253°C
White
Solid
H2KO4P
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
[K+].OP(O)([O-])=O
136.08
CHEBI:63036
≥98.0% (FCC)
Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF)
Poly Bottle

Chemical Identifiers

7778-77-0
136.08
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
potassium dihydrogen phosphate
H2KO4P
MFCD00011401 MFCD00147253
potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
CHEBI:63036
[K+].OP(O)([O-])=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1

SDS Documents

Documents

Promotions

Promotions