missing translation for 'onlineSavingsMsg'
Learn More

Potassium Phosphate Monobasic (Crystalline/NF/FCC), Fisher Chemical™

Catalog No. p-33167
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P380500 500 g Poly Bottle
P380212 2.5 kg Poly Bottle
P38012 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P380500 Supplier Thermo Fisher Scientific Supplier No. FLP380 500
Only null left

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253°C
Color White
pH 4.5
Physical Form Solid
Quantity 500 g
Formula Weight 136.09
Percent Purity 98.0 to 100.5% (NF)
Grade FCC/NF
Identification Pass Test (FCC), Pass Test (FCC/NF), Pass Test (NF)
Loss on Drying 1.0% max. (NF/FCC)
Packaging Poly Bottle
Decomposition Information >250°C
Chemical Name or Material Potassium Phosphate Monobasic
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1