missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate Monobasic, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
P1380, 7778-77-0, KH2PO4
$283.39 - $10117.25
Chemical Identifiers
| CAS | 7778-77-0 |
|---|---|
| Molecular Formula | H2KO4P |
| Molecular Weight (g/mol) | 136.08 |
| MDL Number | MFCD00011401 MFCD00147253 |
| InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
| IUPAC Name | potassium dihydrogen phosphate |
| SMILES | [K+].OP(O)([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605672
|
Spectrum Chemical Mfg Cor
P1380125GM |
125 g | Poly Bottle |
Each for $283.39
|
|
||||
|
18605673
|
Spectrum Chemical Mfg Cor
P1380500GM |
500 g | Poly Bottle |
Each for $585.50
|
|
||||
|
18605675
|
Spectrum Chemical Mfg Cor
P138025KG |
2.5 kg | Poly Bottle |
Each for $1,902.54
|
|
||||
|
18605674
|
Spectrum Chemical Mfg Cor
P1380500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $2,452.54
|
|
||||
|
18605676
|
Spectrum Chemical Mfg Cor
P138025KGCS4 |
4 x 2.5 kg | Poly Bottle |
Case of 4 Each for $6,352.36
|
|
||||
|
18605677
|
Spectrum Chemical Mfg Cor
P138012KG |
12 kg | Poly Pail |
Each for $5,331.86
|
|
||||
|
18605678
|
Spectrum Chemical Mfg Cor
P138050KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Potassium Phosphate Monobasic, Crystal, Reagent, ACS is a soluble salt that is commonly used as a buffering agent. As an ACS grade Reagent, Spectrum Chemical manufactured potassium phosphate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.Chemical Identifiers
| 7778-77-0 | |
| 136.08 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| [K+].OP(O)([O-])=O |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate |
Specifications
| 7778-77-0 | |
| 4.1 to 4.5 | |
| H2KO4P | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| potassium dihydrogen phosphate | |
| 99% | |
| 0.002 |
| 1 | |
| 125 g | |
| MFCD00011401 MFCD00147253 | |
| [K+].OP(O)([O-])=O | |
| 136.08 | |
| ACS | |
| Poly Bottle |