missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Phosphate Monobasic, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
P1380, 7778-77-0, KH2PO4
Supplier: Spectrum Chemical Mfg Cor P1380500GMCS6
Description
Spectrum™ Chemical Potassium Phosphate Monobasic, Crystal, Reagent, ACS is a soluble salt that is commonly used as a buffering agent. As an ACS grade Reagent, Spectrum Chemical manufactured potassium phosphate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.Specifications
| 7778-77-0 | |
| 4.1 to 4.5 | |
| H2KO4P | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| potassium dihydrogen phosphate | |
| 99% | |
| Poly Bottle |
| 1 | |
| 6 x 500 g | |
| MFCD00011401 MFCD00147253 | |
| [K+].OP(O)([O-])=O | |
| 136.08 | |
| 0.002 |
Chemical Identifiers
| 7778-77-0 | |
| 136.08 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| [K+].OP(O)([O-])=O |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate |