missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium phosphate monobasic, ACS Reagent, Honeywell Fluka™
ACS Reagent,≥99.0%
$263.95 - $2409.28
Chemical Identifiers
| CAS | 7778-77-0 |
|---|---|
| Molecular Formula | H2KO4P |
| Molecular Weight (g/mol) | 136.08 |
| MDL Number | MFCD00011401 MFCD00147253 |
| InChI Key | GNSKLFRGEWLPPA-UHFFFAOYSA-M |
| Synonym | potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate |
| PubChem CID | 516951 |
| ChEBI | CHEBI:63036 |
| IUPAC Name | potassium;dihydrogen phosphate |
| SMILES | [K+].OP(O)([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6003141
|
Honeywell-Fluka
P0662500G |
500 g |
Each for $263.95
|
|
|||||
|
6003137
|
Honeywell-Fluka
P06621KG |
1 kg |
Each for $470.95
|
|
|||||
|
6003138
|
Honeywell-Fluka
P066225KG |
2.5 kg |
Each for $726.80
|
|
|||||
|
6003142
|
Honeywell-Fluka
P06626X500G |
6 x 500 g |
Case of 6 Each for $807.85
|
|
|||||
|
6003140
|
Honeywell-Fluka
P06624X25KG |
4 x 2.5 kg |
Case of 4 Each for $2,409.28
|
|
|||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7778-77-0 | |
| 136.08 | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| 516951 | |
| potassium;dihydrogen phosphate |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
| CHEBI:63036 | |
| [K+].OP(O)([O-])=O |
Specifications
| 7778-77-0 | |
| H2KO4P | |
| MFCD00011401 MFCD00147253 | |
| potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate | |
| [K+].OP(O)([O-])=O | |
| 136.08 | |
| CHEBI:63036 | |
| ≥99% | |
| 2.338g/mL at 25°C |
| 500 g | |
| KH2PO4 | |
| NONH for all modes of transport | |
| GNSKLFRGEWLPPA-UHFFFAOYSA-M | |
| potassium;dihydrogen phosphate | |
| 516951 | |
| 136.09g/mol | |
| ACS Reagent | |
| Potassium phosphate monobasic |
Safety and Handling
RTECSNumber : TC6615500