missing translation for 'onlineSavingsMsg'
Learn More

Potassium Oxalate, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

$6800.89 - $6800.89

Chemical Identifiers

CAS 6487-48-5
Molecular Formula C2H2K2O5
Molecular Weight (g/mol) 184.23
MDL Number MFCD00150033
InChI Key QCPTVXCMROGZOL-UHFFFAOYSA-L
Synonym potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
PubChem CID 2724193
IUPAC Name dipotassium hydrate oxalate
SMILES O.[K+].[K+].[O-]C(=O)C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
02003789
View Documents
Avantor J.T.Baker
321205
2.5 kg Wide Mouth Amber Glass Bottle
Each for $6,800.89
Case of 4 Each for $27,203.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6487-48-5
184.23
QCPTVXCMROGZOL-UHFFFAOYSA-L
2724193
O.[K+].[K+].[O-]C(=O)C([O-])=O
C2H2K2O5
MFCD00150033
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
dipotassium hydrate oxalate

Specifications

6487-48-5
2.5 kg
MFCD00150033
QCPTVXCMROGZOL-UHFFFAOYSA-L
dipotassium hydrate oxalate
2724193
BAKER ANALYZED™ ACS Reagent
1L = 2.13kg
Crystals
C2H2K2O5
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
O.[K+].[K+].[O-]C(=O)C([O-])=O
184.23
184.23
Wide Mouth Amber Glass Bottle
Potassium Oxalate, Monohydrate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'