missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrate, Saturated, Ricca Chemical

Catalog No. r6317700100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6317700100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R6317700100 Supplier Ricca Chemical Company Supplier No. R6317700100A

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-79-1 , 7732-18-5
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Color Colorless
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
pH ∼7
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Potassium Nitrate, Saturated
ShelfLife 24 months