missing translation for 'onlineSavingsMsg'
Learn More

Potassium Nitrate, 2.0 M, Ricca Chemical

Catalog No. R6319600120
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6319600120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R6319600120 Supplier Ricca Chemical Company Supplier No. R6319600120A

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7757-79-1 , 7732-18-5
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
Synonym potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Color Colorless
Grade Laboratory
Odor Odorless
Packaging Natural Poly Bottle
pH ∼7
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Potassium Nitrate, 2.0 Molar
ShelfLife 18 months