missing translation for 'onlineSavingsMsg'
Learn More

Potassium Metabisulfite, Crystal, 96%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P1343500GM

 View more versions of this product

Catalog No. 18612786


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

16731-55-8
500 g
Reagent
1
96%
Poly Bottle

Chemical Identifiers

16731-55-8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.