missing translation for 'onlineSavingsMsg'
Learn More

Potassium Metabisulfite, Crystal, 96%, Spectrum™ Chemical

Catalog No. p-7166069
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612785 125 g Poly Bottle
18612786 500 g Poly Bottle
18612787 2.5 kg Poly Pail
18612788 12 kg Poly Pail
18612789 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18612785 Supplier Spectrum Chemical Mfg Cor Supplier No. P1343125GM
Only null left

Chemical Identifiers

CAS 16731-55-8

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 96%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.