missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium iodide, Puriss., meets analytical spec. of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™
Puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Supplier: Honeywell-Fluka 031245KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Potassium Iodide | |
| 7681-11-0 | |
| IK | |
| MFCD00011405 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| 99 to 100.5% (calc. to the dried substance) | |
| Plastic Bottle |
| Meets analytical specification of Ph.Eur. BP, USP | |
| 5 kg | |
| KI | |
| NONH for all modes of transport | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166g/mol | |
| puriss. | |
| 1mmHg (745°C) |
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
| IK | |
| MFCD00011405 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
Safety and Handling
P305 + P351 + P338
H302-H315-H319
EINECSNumber : 231-659-4
RTECSNumber : TT2975000