missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, Puriss., meets analytical spec. of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™

Catalog No. p-7071705
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
6 x 250 g
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001809 250 g
6001808 1 kg
60044510 6 x 250 g
6001810 5 kg
60044509 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6001809 Supplier Honeywell Chemicals Supplier No. 03124250G
Only null left

Puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Quantity 250 g
Linear Formula KI
UN Number NONH for all modes of transport
Formula Weight 166g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 1mmHg (745°C)
Chemical Name or Material Potassium Iodide, Meets analytical specification of Ph.Eur. BP, USP
Hazard Statement P305 + P351 + P338
Precautionary Statement H302-H315-H319
EINECSNumber 231-659-4
RTECSNumber TT2975000