missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, Crystalline, MilliporeSigma™

Catalog No. p-3176448
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX15051 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX15051 Supplier Technidata Supplier No. PX15051
Only null left

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.00
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium iodide
SMILES [K+].[I-]

Specifications

Physical Form Crystalline
Quantity 500 g
Linear Formula KI
Formula Weight 166
Grade ACS
Packaging Poly Bottle
Chemical Name or Material Potassium Iodide