missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, Certified, 10.0% (w/v) ±0.4% (w/v), LabChem™
$68.20 - $1517.53
Chemical Identifiers
| CAS | 7681-11-0 |
|---|---|
| Molecular Formula | IK |
| Molecular Weight (g/mol) | 166.003 |
| InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
| Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
| PubChem CID | 4875 |
| ChEBI | CHEBI:8346 |
| IUPAC Name | potassium;iodide |
| SMILES | [K+].[I-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
LC197601
|
LabChem
LC197601 |
500 mL |
Each for $68.20
|
|
|||||
|
LC197602
|
LabChem
LC197602 |
1 L |
Each for $108.73
|
|
|||||
|
LC197604
|
LabChem
LC197604 |
4 L |
Each for $323.53
|
|
|||||
|
LC197606
|
LabChem
LC197606 |
10 L |
Each for $785.07
|
|
|||||
|
LC197605
|
LabChem
LC197605 |
20 L |
Each for $1,517.53
|
|
|||||
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
Specifications
| 7681-11-0,7732-18-5 | |
| 90,10 | |
| 500 mL | |
| KI | |
| Soluble in water | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| 10.0% ±0.4% (w/v) | |
| Passes Test | |
| 1.07g/mL | |
| 1.07g/mL |
| Yellow | |
| Liquid | |
| IK | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166 | |
| Certified | |
| Amber Poly | |
| Iodine vapor | |
| Potassium Iodide |
Safety and Handling
GHS H Statement
Causes eye irritation.
GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature; Protect from light