missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, Certified, 10.0% (w/v) ±0.4% (w/v), LabChem™
Supplier: LabChem LC197601
Specifications
| Potassium Iodide | |
| Yellow | |
| Liquid | |
| IK | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166 | |
| Certified | |
| Amber Poly | |
| Iodine vapor |
| 7681-11-0,7732-18-5 | |
| 90,10 | |
| 500 mL | |
| KI | |
| Soluble in water | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| 10.0% ±0.4% (w/v) | |
| Passes Test | |
| 1.07g/mL | |
| 1.07g/mL |
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
Safety and Handling
GHS H Statement
Causes eye irritation.
GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature; Protect from light