missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, Certified, 1.00N ±0.01N (1.0M), LabChem™

Catalog No. LC198151
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC198151 500 mL
LC198152 1 L
LC198154 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC198151 Supplier LabChem Supplier No. LC198151

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Color Yellow
CAS Max % 85.15, 14.85
Physical Form Liquid
Quantity 500 mL
Linear Formula KI
Solubility Information Soluble in water
Formula Weight 166
Grade Certified
Concentration or Composition (by Analyte or Components) 1.00N ±0.01N (1.0M)
Identification Passes Test
Packaging Amber Poly
Specific Gravity 1.11g/mL
Decomposition Information Iodine vapor
Density 1.11g/mL
Chemical Name or Material Potassium Iodide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes eye irritation.
Precautionary Statement GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature; Protect from light