missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, ACS Reagent, Honeywell Fluka™
ACS Reagent,≥99.0%
Supplier: Honeywell Chemicals 22194512KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Potassium Iodide | |
| 12 kg | |
| KI | |
| NONH for all modes of transport | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166g/mol | |
| ACS Reagent |
| 7681-11-0 | |
| IK | |
| MFCD00011405 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| ≥99% | |
| 1mmHg (745°C) |
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
| IK | |
| MFCD00011405 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
Safety and Handling
P305 + P351 + P338
H302-H315-H319
EINECSNumber : 231-659-4
RTECSNumber : TT2975000