missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, ACS Reagent, Honeywell Fluka™

Catalog No. p-7071899
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002134 100 g
6002138 500 g
6002136 2.5 kg
6002135 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6002134 Supplier Honeywell Chemicals Supplier No. 221945100G
Only null left

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Quantity 100 g
Linear Formula KI
UN Number NONH for all modes of transport
Formula Weight 166g/mol
Percent Purity ≥99%
Grade ACS Reagent
Vapor Pressure 1mmHg (745°C)
Chemical Name or Material Potassium Iodide
Hazard Statement P305 + P351 + P338
Precautionary Statement H302-H315-H319
EINECSNumber 231-659-4
RTECSNumber TT2975000