missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, (ACS Reagent Grade), MP Biomedicals
$180.56 - $444.64
Chemical Identifiers
| CAS | 7681-11-0 |
|---|---|
| Molecular Formula | IK |
| Molecular Weight (g/mol) | 166.003 |
| InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
| Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
| PubChem CID | 4875 |
| ChEBI | CHEBI:8346 |
| IUPAC Name | potassium;iodide |
| SMILES | [K+].[I-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15256280
|
MP Biomedicals Inc
0215256280 |
100 g |
Each for $180.56
|
|
|||||
|
ICN15256290
|
MP Biomedicals Inc
0215256290 |
500 g |
Each for $444.64
|
|
|||||
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
Specifications
| 7681-11-0 | |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166 | |
| Potassium Iodide |
| 100 g | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| ACS Reagent |
Safety and Handling
Recommended Storage : Room Temperature