missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, (ACS Reagent Grade), MP Biomedicals

Supplier:  MP Biomedicals Inc 0215256290

 View more versions of this product

Catalog No. ICN15256290


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Iodide
500 g
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
[K+].[I-]
166.003
CHEBI:8346
ACS Reagent
7681-11-0
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
166

Chemical Identifiers

7681-11-0
166.003
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]
IK
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions