missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, ACS Grade, ≥ 99.0%, LabChem™
$159.07 - $394.40
Chemical Identifiers
| CAS | 7681-11-0 |
|---|---|
| Molecular Formula | IK |
| Molecular Weight (g/mol) | 166.003 |
| InChI Key | NLKNQRATVPKPDG-UHFFFAOYSA-M |
| Synonym | potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide |
| PubChem CID | 4875 |
| ChEBI | CHEBI:8346 |
| IUPAC Name | potassium;iodide |
| SMILES | [K+].[I-] |
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
Specifications
| 1,330°C | |
| 7681-11-0 | |
| 100 | |
| 100 g | |
| KI | |
| Soluble in water | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| ≥99.0% | |
| Passes Test | |
| 3.13g/cm3 | |
| 3.13g/cm3 |
| 680°C | |
| White | |
| Crystals | |
| IK | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166 | |
| ACS | |
| Amber Glass | |
| Potassium oxide; Iodine vapor | |
| Potassium Iodide |
Safety and Handling
GHS H Statement
Causes eye irritation.
GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 231-659-4
Recommended Storage : Room Temperature; Protect from light