missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, ACS Grade, ≥ 99.0%, LabChem™
Supplier: LabChem LC196901
Specifications
| 1330°C | |
| 680°C | |
| White | |
| Solid | |
| IK | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| potassium;iodide | |
| 4875 | |
| 166 | |
| ACS | |
| Amber Glass | |
| Potassium oxide; Iodine vapor |
| Potassium Iodide | |
| 7681-11-0 | |
| 100 | |
| 500 g | |
| KI | |
| Soluble in water | |
| [K+].[I-] | |
| 166.003 | |
| CHEBI:8346 | |
| ≥99.0% | |
| Passes Test | |
| 3.13g/cm3 | |
| 3.13g/cm3 |
Chemical Identifiers
| 7681-11-0 | |
| 166.003 | |
| potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
| CHEBI:8346 | |
| [K+].[I-] |
| IK | |
| NLKNQRATVPKPDG-UHFFFAOYSA-M | |
| 4875 | |
| potassium;iodide |
Safety and Handling
GHS H Statement
Causes eye irritation.
GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical
Warning
EINECSNumber : 231-659-4
Recommended Storage : Room Temperature; Protect from light