missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodide, 15% w/v, Reagents
Specifications
Specifications
| Boiling Point | 100°C |
| Color | Colorless to Light Yellow |
| Physical Form | Liquid |
| Quantity | 10 L |
| Percent Purity | 15% |
| Grade | Reagent |
| Packaging | Cubitainer |
| Specific Gravity | 1.1086 |
| Chemical Name or Material | Potassium Iodide, 15% w/v |