missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodide, 10% (w/v) Aqueous Solution, Spectrum™ Chemical

Catalog No. 18612759
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612759 500 mL Amber Glass Bottle
18612760 1 L Amber Glass Bottle
18612761 4 L Poly Bottle
18612762 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612759 Supplier Spectrum Chemical Mfg Cor Supplier No. P432500MLGL
Only null left

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.