missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate Volumetric Solution, 0.0500 M (M/20), 0.300 N, Ricca Chemical
$204.77 - $288.33
Chemical Identifiers
| CAS | 7758-05-6 |
|---|---|
| Molecular Formula | IKO3 |
| Molecular Weight (g/mol) | 214.00 |
| InChI Key | JLKDVMWYMMLWTI-UHFFFAOYSA-M |
| Synonym | potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat |
| PubChem CID | 23665710 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R62770-500C
|
Ricca Chemical Company
627716 |
500 mL |
Each for $204.77
|
|
|||||
|
6277-32
|
Ricca Chemical Company
R62770001C |
1 L |
Each for $288.33
|
|
|||||
Chemical Identifiers
| 7758-05-6 | |
| 214.00 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat |
| IKO3 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| 23665710 |
Specifications
| 7758-05-6 , 7732-18-5 | |
| 100.0,1.13 | |
| potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat | |
| [O-]I(=O)=O.[K+] | |
| 214.00 | |
| USP | |
| 500 mL |
| 97.0,1.09 | |
| IKO3 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| potassium;iodate | |
| 23665710 | |
| Amber Glass Bottle | |
| POTASSIUM IODATE VOLUMETRIC SOLUTION |