missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate, 0.125 N, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 98.0, 0.46 |
| CAS Max % | 100.0, 0.48 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 0.125 Normal |
| Packaging | Amber Glass Bottle |
| Chemical Name or Material | Potassium Iodate |
Safety and Handling
| ShelfLife | 18 months |