missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, 0.125 N, Ricca Chemical

Catalog No. 6276532
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6276532 1 L Amber Glass Bottle
627651 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6276532 Supplier Ricca Chemical Company Supplier No. 6276.532
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7758-05-6
Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
Synonym potassium iodate, potassium triodate, caswell no. 693a, iodic acid, potassium salt, unii-i139e44nhl, potassium iodine oxide kio3, iodic acid hio3 , potassium salt, epa pesticide chemical code 075703, iodic acid hio3 , potassium salt 1:1, kaliumjodat
PubChem CID 23665710
IUPAC Name potassium;iodate
SMILES [O-]I(=O)=O.[K+]

Specifications

Color Colorless
CAS Min % 98.0, 0.46
CAS Max % 100.0, 0.48
Physical Form Liquid
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.125 Normal
Packaging Amber Glass Bottle
Chemical Name or Material Potassium Iodate
ShelfLife 18 months