missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Iodate, 0.0250 N (N/40), Ricca Chemical
ASTM for Sulfite Testing
$196.53 - $196.53
Chemical Identifiers
| Molecular Formula | IKO3 |
|---|---|
| Molecular Weight (g/mol) | 214.00 |
| InChI Key | JLKDVMWYMMLWTI-UHFFFAOYSA-M |
| IUPAC Name | potassium iodate |
| SMILES | [K+].[O-][I](=O)=O |
Chemical Identifiers
| IKO3 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| [K+].[O-][I](=O)=O |
| 214.00 | |
| potassium iodate |
Specifications
| 5-6-7758 | |
| JLKDVMWYMMLWTI-UHFFFAOYSA-M | |
| potassium iodate | |
| Laboratory | |
| 1 L |
| IKO3 | |
| [K+].[O-][I](=O)=O | |
| 214.00 | |
| Amber Glass Bottle | |
| Potassium Iodate |