missing translation for 'onlineSavingsMsg'
Learn More

Potassium Iodate, 0.0250 N (N/40), Ricca Chemical

Catalog No. 627332
Change view
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
627332 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 627332 Supplier Ricca Chemical Company Supplier No. 627332

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ASTM for Sulfite Testing

Chemical Identifiers

Molecular Formula IKO3
Molecular Weight (g/mol) 214.00
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
IUPAC Name potassium iodate
SMILES [K+].[O-][I](=O)=O

Specifications

CAS 5-6-7758
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 1 L
Chemical Name or Material Potassium Iodate