missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, in Ethanol, Certified, 0.500N ±0.005N (0.5M) , LabChem™

Catalog No. LC195602
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC195602 1 L Poly Bottle
LC195604 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC195602 Supplier LabChem Supplier No. LC195602
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1310-58-3 | HKO | 56.11 g/mol

Chemical Identifiers

CAS 1310-58-3 , 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless to Orange
CAS Max % 2.76, 88, 4.863, 4.377
Linear Formula KOH
Concentration 0.500N ± 0.005N
UN Number UN2924
Solubility Information Soluble in water
Quantity 1 L
Formula Weight 56.11
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.5N (0.5M) in Ethanol, in Ethanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
May damage fertility or the unborn child (oral).
Causes damage to organs (cen
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Recommended Storage Room Temperature