missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, in Ethanol, Certified, 0.100N ±0.001N (0.1M), LabChem™

Catalog No. LC193102
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC193102 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC193102 Supplier LabChem Supplier No. LC193102
Only null left

Chemical Identifiers

CAS 1310-58-3 , 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color Colorless
CAS Max % 0.6, 89.96, 4.97, 4.47
Linear Formula KOH
Concentration 0.100N ±0.001N (0.1M)
UN Number UN1170
Solubility Information Soluble in water
Quantity 1 L
Formula Weight 56.11
Grade Certified
Identification Passes Test
Packaging Amber Glass
Traceability to NIST Traceable to NIST
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, in Ethanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve) (oral, dermal).
Precautionary Statement GHS P Statement
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
If swallowed: Rinse mouth.
Call a poison center/doctor.
In case of fire: Use carbon dioxide (CO2), powder, alcohol-resistant foam to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature