missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 2.00 Normal, Reagents
Potassium Hydroxide, 2.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CP6000004A
Specifications
| 14 | |
| Colorless | |
| 7732-18-5,1310-58-3 | |
| Caustic Potash, Potash Lye | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| 1.0913 |
| Potassium Hydroxide, 2.00 Normal | |
| 100°C | |
| H2O | |
| 4 L | |
| O | |
| 18.02 | |
| Poly Bottle | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Caustic Potash, Potash Lye | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |