missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 2.00 Normal, Reagents

Catalog No. p-200021964
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334093 1 L
01334092 20 L
01334091 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334093 Supplier Labchem Inc Supplier No. CP6000001A
Only null left

Potassium Hydroxide, 2.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1310-58-3
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name water
SMILES O

Specifications

pH 14
Color Colorless
Boiling Point 100°C
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0913
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 2.00 Normal