missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.500 N (N/2) in Methanol, Ricca Chemical

$212.20 - $1535.43

Chemical Identifiers

CAS 67-56-1
Molecular Formula HKO
Molecular Weight (g/mol) 56.11 g/mol
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
626116
View Documents
Ricca Chemical Company
626116
500 mL Natural Poly Bottle
Each for $212.20
Only null left
 
R62610004F
View Documents
Ricca Chemical Company
R62610004F
4 L Cubitainer
Each for $620.40
Only null left
 
62612-5
View Documents
Ricca Chemical Company
626125
10 L Cubitainer
Each for $1,535.43
Only null left
 
Description

Description

  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability
Specifications

Chemical Identifiers

67-56-1
56.11 g/mol
KWYUFKZDYYNOTN-UHFFFAOYSA-M
[OH-].[K+]
HKO
MFCD00003553
potassium hydroxide

Specifications

Colorless
3.86,94%
HKO
0.5 Normal
500 mL
[OH-].[K+]
56.11 g/mol
Natural Poly Bottle
Liquid
67-56-1,1310-58-3,7732-18-5
4.02,98%
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, columbian spirit
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
Laboratory
Methanol
Potassium Hydroxide
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'