missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 0.500 N (N/2) in Methanol, Ricca Chemical
Supplier: Ricca Chemical Company 626125
Description
- Tested in an ISO 17025 Accredited Facility
- Test Reports provided with Certified Values and Measurement Uncertainty
- Tight release specifications for low lot-to-lot variability
Specifications
| Potassium Hydroxide | |
| 67-56-1,1310-58-3,7732-18-5 | |
| 4.02,98% | |
| MFCD00003553 | |
| potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, columbian spirit | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| potassium hydroxide | |
| Laboratory | |
| Methanol |
| Colorless | |
| 3.86,94% | |
| HKO | |
| 0.500Normal | |
| 10 L | |
| [OH-].[K+] | |
| 56.11 g/mol | |
| Cubitainer | |
| Liquid |
Chemical Identifiers
| 67-56-1 | |
| 56.11 g/mol | |
| KWYUFKZDYYNOTN-UHFFFAOYSA-M | |
| [OH-].[K+] |
| HKO | |
| MFCD00003553 | |
| potassium hydroxide |