missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydroxide, 0.500 N (N/2) in Methanol, Ricca Chemical
Description
- Tested in an ISO 17025 Accredited Facility
- Test Reports provided with Certified Values and Measurement Uncertainty
- Tight release specifications for low lot-to-lot variability
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 3.86, 94% |
| CAS Max % | 4.02, 98% |
| Concentration or Composition Notes | 0.5 Normal |
| Synonym | potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, columbian spirit |
| Quantity | 500 mL |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Methanol |
| Physical Form | Liquid |
| Show More |