missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.500 N in Denatured Ethanol (Reagent Alcohol), Reagents

Catalog No. 01334469
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01334469 1 L Poly Bottle
01334467 20 L Cubitainer
01334466 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01334469 Supplier Labchem Inc Supplier No. CP1014001A
Only null left

Potassium Hydroxide, 0.500 N in Denatured Ethanol (Reagent Alcohol), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name ethanol
SMILES CCO

Specifications

pH >13
Color Colorless
Boiling Point 78°C
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.8168
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.500 N in Denatured Ethanol (Reagent Alcohol)