missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.100 N (N/10), Ricca Chemical

$121.30 - $1076.40

Chemical Identifiers

CAS 1310-58-3
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6200-16
SDS Accessories View Documents
Ricca Chemical Company
620016
500 mL Natural Poly Bottle
Each for $121.30
Only null left
 
6200-32
SDS Accessories View Documents
Ricca Chemical Company
620032
1 L Natural Poly Bottle
Each for $142.87
Only null left
 
6200-1
Accessories View Documents
Ricca Chemical Company
RI62001
4 L Natural Poly Bottle
Each for $483.50
Only null left
 
6200-5
View Documents
Ricca Chemical Company
RI62005
20 L Cubitainer
Each for $1,076.40
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]

Specifications

1310-58-3 , 7732-18-5
100.0,0.65
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
Laboratory
13
0.100N
97.0,0.63
HKO
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]
56.11
CHEBI:32035
Natural Poly Bottle
500 mL
Potassium Hydroxide
Videos
SDS
Documents

Documents