missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.050 Normal (N/20) in Reagent Alcohol, Reagents

Catalog No. 01334383
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334383 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334383 Supplier Labchem Inc Supplier No. CP1091001A
Only null left

Potassium Hydroxide, 0.050 Normal (N/20) in Reagent Alcohol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Caustic Potash, Potash Lye
IUPAC Name ethanol
SMILES CCO

Specifications

pH >12
Color Colorless
Boiling Point 78°C
Quantity 1 L
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.8025
Physical Form Liquid
Chemical Name or Material Potassium Hydroxide, 0.050 Normal (N/20) in Reagent Alcohol