missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, 0.0200 N (N/50), Ricca Chemical

Supplier:  Ricca Chemical Company 618732

 View more versions of this product

Catalog No. 618732


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Hydroxide
1310-58-3 , 7732-18-5
100.0,0.26
MFCD00003553
KWYUFKZDYYNOTN-UHFFFAOYSA-M
potassium hydroxide
14797
Laboratory
>12
Liquid
Colorless
98.0,0.25
HKO
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
[OH-].[K+]
56.11
CHEBI:32035
Natural Poly Bottle
1 L
0.02 N

Chemical Identifiers

1310-58-3
56.11
KWYUFKZDYYNOTN-UHFFFAOYSA-M
14797
potassium hydroxide
HKO
MFCD00003553
potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
CHEBI:32035
[OH-].[K+]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions