missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Gluconate, Anhydrous, FCC, 98%, Spectrum™ Chemical
P1314, 299-27-4, C6H11KO7
Supplier: Spectrum Chemical Mfg Cor P131425KG
Description
Spectrum™ Chemical Potassium Gluconate, Anhydrous, FCC is used as a mineral supplement and sequestrant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 299-27-4 | |
| 2.5 kg | |
| HLCFGWHYROZGBI-JJKGCWMISA-M | |
| potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate | |
| 98% | |
| 0.03 |
| 1 | |
| C6H11KO7 | |
| [K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O | |
| 234.25 | |
| FCC | |
| Poly Pail |
Chemical Identifiers
| 299-27-4 | |
| 234.25 | |
| potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate |
| C6H11KO7 | |
| HLCFGWHYROZGBI-JJKGCWMISA-M | |
| [K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O |