missing translation for 'onlineSavingsMsg'
Learn More

Potassium Gluconate, Anhydrous, FCC, 98%, Spectrum™ Chemical

Catalog No. p-7157942
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605572 500 g Poly Bottle
18605573 2.5 kg Poly Pail
18605574 12 kg Poly Pail
18605575 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605572 Supplier Spectrum Chemical Mfg Cor Supplier No. P1314500GM
Only null left

P1314, 299-27-4, C6H11KO7

Spectrum™ Chemical Potassium Gluconate, Anhydrous, FCC is used as a mineral supplement and sequestrant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 299-27-4
Molecular Formula C6H11KO7
Molecular Weight (g/mol) 234.25
InChI Key HLCFGWHYROZGBI-JJKGCWMISA-M
IUPAC Name potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES [K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 98%
Grade FCC
Loss on Drying 0.03
Packaging Poly Bottle