missing translation for 'onlineSavingsMsg'
Learn More

Potassium formate, 96%, pract.

CAS: 590-29-4 | CHKO2 | 84.12 g/mol

Supplier:  Thermo Scientific Chemicals 418225000

 View more versions of this product

Catalog No. AC418225000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium formate
165°C to 168°C
Authentic
Plastic bottle
HCOOK
500 g
potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
WFIZEGIEIOHZCP-UHFFFAOYSA-M
potassium;formate
2735122
84.12
Practical
590-29-4
White
96%
CHKO2
MFCD00013100
15,7754
Solubility in water: soluble in water
C(=O)[O-].[K+]
84.12
CHEBI:63316
96%
Crystalline Powder, Crystals or Chunks

Chemical Identifiers

590-29-4
84.12
WFIZEGIEIOHZCP-UHFFFAOYSA-M
2735122
potassium;formate
CHKO2
MFCD00013100
potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
CHEBI:63316
C(=O)[O-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-677-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only