missing translation for 'onlineSavingsMsg'
Learn More

Potassium formate, 96%, pract.

Catalog No. p-3737247
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Plastic bottle
Plastic Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC418221000 100 g Plastic bottle
AC418225000 500 g Plastic bottle
AC418220050 5 kg Plastic Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC418221000 Supplier Thermo Scientific Chemicals Supplier No. 418221000
Only null left

CAS: 590-29-4 | CHKO2 | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 590-29-4
Molecular Formula CHKO2
Molecular Weight (g/mol) 84.12
MDL Number MFCD00013100
InChI Key WFIZEGIEIOHZCP-UHFFFAOYSA-M
Synonym potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
PubChem CID 2735122
ChEBI CHEBI:63316
IUPAC Name potassium;formate
SMILES C(=O)[O-].[K+]

Specifications

Melting Point 165°C to 168°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic bottle
Linear Formula HCOOK
Quantity 100 g
Merck Index 15,7754
Solubility Information Solubility in water: soluble in water
Formula Weight 84.12
Percent Purity 96%
Grade Practical
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Potassium formate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-677-9

RUO – Research Use Only