missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Fluoride, purum p.a., ≥99.0% (F), Honeywell™
Purum p.a.,≥99.0% (F)
$406.76 - $909.95
Chemical Identifiers
| CAS | 7789-23-3 |
|---|---|
| Molecular Formula | FK |
| Molecular Weight (g/mol) | 58.10 |
| MDL Number | MFCD00011398 |
| InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
| Synonym | potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 |
| PubChem CID | 522689 |
| ChEBI | CHEBI:66872 |
| IUPAC Name | potassium;fluoride |
| SMILES | [F-].[K+] |
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
| 7789-23-3 | |
| 58.10 | |
| NROKBHXJSPEDAR-UHFFFAOYSA-M | |
| 522689 | |
| potassium;fluoride |
| FK | |
| MFCD00011398 | |
| potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
| CHEBI:66872 | |
| [F-].[K+] |
Specifications
| 7789-23-3 | |
| FK | |
| MFCD00011398 | |
| 3902818 | |
| NROKBHXJSPEDAR-UHFFFAOYSA-M | |
| potassium;fluoride | |
| 522689 | |
| 58.1g/mol | |
| Potassium fluoride |
| 250 g | |
| KF | |
| UN1812 | |
| potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
| [F-].[K+] | |
| 58.10 | |
| CHEBI:66872 | |
| ≥99% (F) |
Safety and Handling
P261-P280-P301 + P310-P311