missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Fluoride, purum p.a., ≥99.0% (F), Honeywell™
Purum p.a.,≥99.0% (F)
Supplier: Honeywell Inc 6024025KG
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
| Potassium fluoride | |
| 2.5 kg | |
| KF | |
| UN1812 | |
| potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
| [F-].[K+] | |
| 58.10 | |
| CHEBI:66872 | |
| ≥99% (F) |
| 7789-23-3 | |
| FK | |
| MFCD00011398 | |
| 3902818 | |
| NROKBHXJSPEDAR-UHFFFAOYSA-M | |
| potassium;fluoride | |
| 522689 | |
| 58.1g/mol |
Chemical Identifiers
| 7789-23-3 | |
| 58.10 | |
| NROKBHXJSPEDAR-UHFFFAOYSA-M | |
| 522689 | |
| potassium;fluoride |
| FK | |
| MFCD00011398 | |
| potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
| CHEBI:66872 | |
| [F-].[K+] |
Safety and Handling
P261-P280-P301 + P310-P311