missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, Dihydrate, Crystal, Reagent, 99-102%, Spectrum™ Chemical

P1310, 13455-21-5, KF·2H2O

$212.47 - $25932.93

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
IUPAC Name potassium dihydrate fluoride
SMILES O.O.[F-].[K+]
View More Specs

Products 7
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18605565
View Documents
Spectrum Chemical Mfg Cor
P131025GM
25 g Poly Bottle
Each for $212.47
Only null left
 
18605566
View Documents
Spectrum Chemical Mfg Cor
P1310125GM
125 g Poly Bottle
Each for $488.13
Only null left
 
18605567
View Documents
Spectrum Chemical Mfg Cor
P1310500GM
500 g Poly Bottle
Each for $1,268.33
Only null left
 
18605569
View Documents
Spectrum Chemical Mfg Cor
P131025KG
2.5 kg Poly Bottle
Each for $4,491.33
Only null left
 
18605568
View Documents
Spectrum Chemical Mfg Cor
P1310500GMCS6
6 x 500 g Poly Bottle
Case of 6 Each for $6,758.57
Only null left
 
18605570
View Documents
Spectrum Chemical Mfg Cor
P131012KG
12 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18605571
View Documents
Spectrum Chemical Mfg Cor
P131045KGBL
45 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Potassium Fluoride, Dihydrate, Crystal, Reagent is a source of fluoride, it is the preferred reagent in the conversion process of chlorocarbons into fluorocarbons. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Specifications

Chemical Identifiers

13455-21-5
94.13
MZFXHSQFQLKKFD-UHFFFAOYSA-M
O.O.[F-].[K+]
FH4KO2
MFCD00149929
potassium dihydrate fluoride

Specifications

13455-21-5
25 g
MFCD00149929
O.O.[F-].[K+]
94.13
Reagent
1
FH4KO2
MZFXHSQFQLKKFD-UHFFFAOYSA-M
potassium dihydrate fluoride
99 to 102%
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'